ContactUsLinkComponent
HBM® Basal Medium
HBM® Basal Medium
Catalog #: CA10065-288
CAS Number:  
Clonetics* HBM Basal Medium 500 ml
HBM® Basal Medium
Catalog #: CA10065-288
CAS Number:  
Specifications
  • Description:
    HBM® Basal medium, 500 ml
  • Cat. no.:
    CA10065-288
Specifications
About this item
The HBM®Basal Medium consists of 500Mls hepatocyte basal medium without supplementation. It is designed to be used only in conjunction with the HCM® SingleQuots® (CC-4182) . Once combinded together these reagents formulate a highly reproducible maintenance medium for primary hepatocytes from all species. The final medium is serum-free.
  • Benefits:
  • Optimized for Hepatocyte Cell Health, Compatible with all species hepatocytes
  • Content:
  • 500 ml
Customers Who Bought This Also Bought