BulletKit® HCM® Hepatocyte Culture Medium
Catalog # CA10065-286
Supplier: LONZA PHARMA - BIOSCIENCE CA
CAS Number:
Specifications
- Description:HCM® Hepatocyte culture medium
- Cat. no.:CA10065-286
Specifications
About this item
The Hepatocyte Culture Medium BulletKit® consists of the HBM® Hepatocyte Basal Medium, 500 ml (CC-3199) and the HCM® SingleQuots® Kit (CC-4182). Once combinded together these reagents formulate a highly reproducible maintenance medium for primary hepatocytes from all species. The final medium is serum-free.
- Optimized for hepatocyte cell health
- Compatible with all species hepatocytes
Delivery information: Content:
1 × HBM® (CC-3199) Bottle Basal Medium, 500 ml
1 × HCM® (CC-4182) Singlequots® Supplement Pack containing:
1 × Transferrin, 0.50 ml
1 × Ascorbic Acid, 0.50 ml
1 × HEGF, 0.50 ml
1 × Insulin, 0.50 ml
1 × Hydrocortisone, 0.50 ml
1 × BSA (Fatty Acid Free), 10.5 ml
1 × GA-1000, 0.50 ml
1 × HBM® (CC-3199) Bottle Basal Medium, 500 ml
1 × HCM® (CC-4182) Singlequots® Supplement Pack containing:
1 × Transferrin, 0.50 ml
1 × Ascorbic Acid, 0.50 ml
1 × HEGF, 0.50 ml
1 × Insulin, 0.50 ml
1 × Hydrocortisone, 0.50 ml
1 × BSA (Fatty Acid Free), 10.5 ml
1 × GA-1000, 0.50 ml