4D-Nucleofector® X SE Cell Line Kit L
Supplier: LONZA PHARMA - BIOSCIENCE CA
Certificates
About this item
The SE Cell Line 4D-Nucleofector® X Kit L is one of our three 4D-Nucleofector® Kits that is suited for transfecting difficult-to-transfect cell lines in the 100 µl Nucleocuvette® Vessel format when working with the 4D-Nucleofector® X Unit.
- High transfection efficiencies in difficult-to-transfect cell lines
- Cells maintain functionality post transfection
- Conditions are transferable between 4D-Nucleofector® System, 96-well Shuttle® System and 384-well Nucleofector® System
Applications: Transfection of cell numbers in the range of 2×10⁵ to 2×10⁷ cells, Low throughput applications.
The kit is available in different variants for either 12 or 24 reactions.
Delivery: Contents:
12 Reactions: 2 × 0.675 ml SE Cell Line Nucleofector® solution, 2 × 0.15 ml Supplement 1, 50 µg pmaxGFP® Vector (1 μg/μl in 10 mM Tris pH 8.0), 12 × 100 µl Nucleocuvette® Vessel, 12 × Single use pipettes.
24 Reactions: 2.25 ml SG Cell Line Nucleofector® solution, 0.5 ml Supplement 1, 50 µg pmaxGFP® Vector (1 μg/μl in 10 mM Tris pH 8.0), 24 × 100 µl Nucleocuvette® Vessel, 24 × Single use pipettes.
12 Reactions: 2 × 0.675 ml SE Cell Line Nucleofector® solution, 2 × 0.15 ml Supplement 1, 50 µg pmaxGFP® Vector (1 μg/μl in 10 mM Tris pH 8.0), 12 × 100 µl Nucleocuvette® Vessel, 12 × Single use pipettes.
24 Reactions: 2.25 ml SG Cell Line Nucleofector® solution, 0.5 ml Supplement 1, 50 µg pmaxGFP® Vector (1 μg/μl in 10 mM Tris pH 8.0), 24 × 100 µl Nucleocuvette® Vessel, 24 × Single use pipettes.
Customers Who Bought This Also Bought