Order Entry
ContactUsLinkComponent
Rat Brain Hippocampus Neurons
Rat Brain Hippocampus Neurons
Catalog #: CA10064-166
CAS Number:  
R-Hi - Rat Brain Hippocampus Neurons, Cryopreserved, in PNGM* BulletKit* =1 million cells/vial
Rat Brain Hippocampus Neurons
Catalog #: CA10064-166
CAS Number:  

Specifications

  • Description:
    Rat brain hippocampus neurons
  • Size:
    ≥1 million cells
  • Cat. no.:
    CA10064-166

Specifications

About this item

Ready-to-use Clonetics® Rat Brain Hippocampus Neurons from rat (E18, 19) brain are cell suspensions of high quality primary embryonic brain neuronal cells (including glia) prepared by standardized methods, and are ready for immediate culture.

  • Ready-To-Use
  • No tissue dissection or animal handling required
  • Increased data reproducibility
  • Characteristic morphology and staining tests confirm tissue origin and cell type
  • Negative for mycoplasma, bacteria, yeast and fungi

Each vial of cells contains approximately 1 million viable cells. This will seed into 5 or more wells of a 24-well plate and 25 or more wells of a 96-well plate using the recommended plating densities and medium. Cell death will occur during the first few days after plating and debris will be observed. This is normal. After approximately 4 days in culture, the cells will form a neurite network and by the 7th day, debris will be minimal.

Recommended Media: PNGM® BulletKit® (CC-4461)

Applications: Neurogenesis, Neurotransmitter function, Gene expression, Signaling pathways, Electrophysiology, Neurotoxicity, Drug/Compound screening, Advanced cell culture models.

Delivery: Content: ≥1000000 cryopreserved cells per ampule. Certificate of Analysis (CoA) is available for every batch.
Customers Who Bought This Also Bought