Specifications
- Description:hADSC Human adipose-derived stem cells
- Size:≥1 million cells
- Cat. no.:CA10063-988
- Supplier No.:PT-5006
Specifications
About this item
Poietics® Normal Human Adipose Derived Stem Cells (hADSC) are isolated from normal (non-diabetic) adult lipoaspirates collected during elective surgical liposuction procedures. Normal Human ADSC have been reported to differentiate down many different lineages including chondrogenic, osteogenic, adipogenic and neural. Normal Human ADSC have been cryopreserved at primary passage.
- All cells test negative for mycoplasma, bacteria, yeast, and fungi
- HIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lots
Pertinent additional donor information, including Body Mass Index (BMI) may be available by contacting Scientific Support. Cryopreserved Poietics® Normal Human ADSC are guaranteed through 5 passages, to express CD13, CD29, CD44, CD73, CD90, CD105, and CD166 and to not express CD14, CD31, and CD45.
Human ADSC from diabetic donors are available as a separate product. These cells have demonstrated very similar phenotypic and functional characteristics to that of bone marrow-derived mesenchymal stem cells.
Recommended Media and Reagents:
Growth Medium:
ADSC Human Adipose Derived Stem Cell Growth BulletKit® Medium
Differentiation Medium*:
Adipogenic: PGM® -2 Preadipocyte Growth Medium-2 BulletKit® Medium (provides both a plating medium and a differentiation medium appropriate to induce differentiation of human adipose derived mesenchymal stem cells into mature, functionally active adipocytes; Each bottle of the PGM-2® BulletKit® Medium contains enough differentiation medium to fully differentiate up to 20 million Adipose Derived Stem Cells (ADSC) [10 complete 96-well plates] into mature, functionally active adipocytes).
Chondrogenic: hMSC Chondrogenic Differentiation BulletKit® Medium, with the addition of TGF-β3 (provides a differentiation medium appropriate to induce chondrogenic differentiation of human adipose derived mesenchymal stem cells into mature, functionally active chondrocytes; Each bottle of the hMSC Chondrogenic Differentiation BulletKit® Medium, with the addition of TGF-β3 (sold separately), contains enough differentiation medium to fully differentiate approximately 12.5 million human adipose derived mesenchymal stem cells [approximately 25 micropellets] into mature, functionally active chondrocytes).
Osteogenic: hMSC Osteogenic Differentiation BulletKit® Medium (provides a differentiation medium appropriate to induce osteogenic differentiation of human adipose derived mesenchymal stem cells into mature, functionally active osteoblasts; Each bottle of the hMSC Osteogenic Differentiation BulletKit® Medium contains enough differentiation medium to fully differentiate approximately 200000 human adipose derived mesenchymal stem cells [approximately 20 wells in a 24-well plate format] into mature, functionally active osteoblasts).