HCAEC – Human Coronary Artery Endothelial Cells
Catalog #: CA10064-274
Supplier: LONZA PHARMA - BIOSCIENCE CA
CAS Number:
Specifications
- Description:HCAEC – Human coronary artery endothelial cells
- Size:≥500000 cells
- Cat. no.:CA10064-274
- Supplier No.:CC-2585
Specifications
About this item
Cardiac Endothelial Cells include cells isolated from the aorta (HAEC), coronary artery (HCAEC), pulmonary artery (HPAEC), iliac artery (HIAEC), and microvascular cardiac cells (HMVEC-C).
- Assured for experimental use up to 15 population doublings
- All cells test negative for mycoplasma, bacteria, yeast, and fungi
- HIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lots
Clonetics® Cells, Medium and Reagents are quality tested together and guaranteed to give optimum performance as a complete cell system. Cryopreserved HCAEC are shipped in third passage.
Cryopreserved Clonetics® Human Coronary Arteryc Endothelial Cells (HCAEC) are guaranteed through 15 population doublings, express CD31/105, von Williebrand Factor VIII, and are positive for acetyated low density lipoprotein uptake.
Applications: Atherosclerosis, Angiogenesis, Cell-to-cell Junctions, Inflammation, Wound Healing, Oncology.
Delivery: Content:1 × Cryopreserved ampule of HCAEC containing ≥500000 cells/vial. A Certificate of Analysis is provided for each cell lot purchased.
Customers Who Bought This Also Bought