ContactUsLinkComponent
HMVEC-dAd – Human Dermal Microvascular Endothelial Cells – Adult
HMVEC-dAd – Human Dermal Microvascular Endothelial Cells – Adult
Catalog #: CA10065-218
CAS Number:  
Clonetics* HMVEC-d Ad-Dermal MV Endo ,EGM-2MV, amp
HMVEC-dAd – Human Dermal Microvascular Endothelial Cells – Adult
Catalog #: CA10065-218
CAS Number:  
Specifications
  • Description:
    HMVEC-dAd – Human dermal microvascular endothelial cells – adult
  • Size:
    ≥500000 cells
  • Cat. no.:
    CA10065-218
  • Supplier No.:
    CC-2543
Specifications
About this item
Human Dermal Microvascular Endothelial Cells include cells isolated from small vessels within skin tissue from either adults or neonatal foreskins.Cells are cultured to be ≥90% pure and specifically enriched for dermal LECs and BECs.
  • Ready-To-Use
  • No tissue dissection required
  • Increased data reproducibility
  • Characteristic morphology and staining tests confirm tissue origin and cell type
  • Negative for mycoplasma, bacteria, yeast and fungi
  • HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots
Dermal microvascular endothelial cells, from either neonates or adults (HMVEC-D) are a mixed population of both blood (HMVEC-DBl) and lymphatic (HMVEC-DLy) endothelial cells.
Adult cells are designated with an 'Ad'; neonate cells are designated with 'Neo'. Both pooled and single donors are available. Endothelial Cells and Media are convenient and easy to use, allowing the researcher to focus on results. Lonza's Cells, Medium and Reagents are quality tested together and guaranteed to give optimum performance as a complete cell system.
Cryopreserved Adult Human Dermal Microvascular Endothelial Cells are shipped in third passage. Cryopreserved Dermal Microvascular Endothelial Cells are guaranteed through 15 population doublings (CC-2543, CC-2505 and CC-2516) and 12 population doublings (CC-2810, CC-2811, CC-2812 and CC-2813) , express CD31/105, von Williebrand Factor VIII, and are positive for acetyated low density lipoprotein uptake.
Delivery: Content: 1 × Cryopreserved Ampule with ≥500000 cells/vial. Certificate of Analysis (CoA) is available for each cell lot purchased.
Customers Who Bought This Also Bought