To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:PRAS40
- Clonality:Polyclonal
- Gene ID:84335
- Host:Rabbit
- Isotype:IgG (H, L)
- Reactivity:Human
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µl
- Western blot:Yes
- Storage buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Molecular weight:28
- Storage temperature:Shipped at 4 ℃. Store at −20 °C for one year. Avoid repeated freeze/thaw cycles.
- Concentration:1 µg/µl
- Purification:Purified by protein A
- Packaging:Vial
Specifications
About this item
Proline Rich Akt Substrate (PRAS40) is a 40 kDa substrate of Akt. Akt and PRAS40 can be found in the phosphoinositide 3 kinase (PI3K) pathway, which plays a role in glucose uptake, cell growth, and apoptosis inhibition. PRAS40 is a 14-3-3 binding protein that reacts with insulin, but whose precise function is not yet known. It may bind SH3 and WW domain containing proteins resulting in a change of function. Activated Akt phosphorylates PRAS40 on threonine 246. Mutation of PRAS40 threonine 246 has been shown to be apoptotic.
Type: Primary
Antigen: PRAS40
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Human