Nucleofector® Primary Cell Optimization 4D-Y Kit
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Electroporation-based methods have so far required cells to be in suspension for transfection. However, the 4D-Nucleofector® Y Unit allows direct Nucleofection of cells in adherence. Cells which typically grow in adherence in culture can be kept and transfected by Nucleofection in their physiological state.
- Convenient and rapid determination of optimal conditions for Nucleofection of cells in adherence within one experiment
The primary cell optimization 4D-Nucleofector® Y kit is suited for rapid determine Nucleofection conditions for primary cell types lacking an Optimized Protocol for the 4D-Nucleofector® Y Unit. Using one 24-well Dipping Electrode Array and our pmaxGFP® Vector as positive control, our two Primary Cell Nucleofector® Solutions for adherent Nucleofection (AD1 and AD2) are tested in combination with a pre-selected set of programs plus controls. The remaining 24-well Dipping Electrode Array can be used for optional fine tuning or performing the first experiment with the selected conditions.
The cells are cultured in the 24-well culture plates provided with the kit*. For the transfection step, a 24-well Dipping Electrode Array is inserted after replacing culture medium by the specific Nucleofector® Solution mixed with the DNA or RNA substrate.
Applications:
Determination of nucleofection conditions for primary cell types lacking an optimized protocol for the 4D-Nucleofector® Y unit.
Delivery information: Content:
2×4.5 ml AD1 4D-Nucleofector® Y solution.
2×4.5 ml AD2 4D-Nucleofector® Y solution.
4×1 ml Supplement 1.
6×150 μg pmaxGFP® vector (1 μg/μl in 10 mM Tris pH 8.0).
2×24-well Dipping Electrode Array.
2×Nunclon® Δ Surface 24-well plate (Nunc).
Specifications
Specifications
- Description:Primary cell 96-well Nucleofector® kit
- No. of reactions:48
Specifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



