Specifications
- Description:S-ALI® Small airway air-liquid interface medium BulletKit®
- Cat. no.:CA10064-040
Specifications
About this item
S-ALI® BulletKit® Small airway air-liquid interface BulletKit® provides scientists with a unique tool to investigate new areas of airway epithelial research in a more physiologically relevant way.
By using an air-liquid interface model, the next generation tool of in vitro respiratory research, scientists can explore small airway epithelial cell differentiation and apply therapies in novel ways. The S-ALI® Bullekit® is made up of a growth and differentiation medium SingleQuots® Kit, growth basal medium and differentiation basal medium.
Our S-ALI® BulletKit® promotes full differentiation of the small airway epithelium expressed by the formation of a polarized epithelium with good barrier function (transepithelial resistance), secretory phenotype (mucin secretion) and ciliogenesis.When used with our guaranteed small airway epithelial cells (CC-2547S), differentiation will occur by day 22 in culture.
1×S-ALI® Growth basal media (CC-3281), 250 ml.
1×S-ALI® Differentiation media (CC-3282), 500 ml.
1×S-ALI® SingleQuots® Supplement pack (CC-4538) containing:
1×Natural Cap vial with transferrin, 0.75 ml.
1×Amber vial with S-ALI-inducer, 1.00 ml.
1×Orange cap vial with BPE, 3.00 ml.
1×Amber vial with epinephrine, 0.75 ml.
1×Red cap vial with GA-1000, 0.75 ml.
1×Green cap vial hEGF, 0.75 ml.
1×Natural cap vial with hydrocortisone, 0.75 ml.
1×Lilac cap vial with insulin, 0.75 ml.
1×Amber vial with retinoic acid, 0.75 ml.
1×Bottle fatty acid free BSA-FAF, 7.50 ml.
1×Amber vial with triiodothyronine, 0.75 ml.