D-HCAEC – Diseased Human Coronary Artery Endothelial Cells – Diabetes Type II
Catalog #: CA10065-222
Supplier: LONZA PHARMA - BIOSCIENCE CA
CAS Number:
Specifications
- Description:D-HCAEC – Diseased Human Coronary Artery Endothelial Cells – diabetes type II
- Size:≥500000 cells
- Cat. no.:CA10065-222
Specifications
About this item
Diseased human coronary artery endothelial cells are isolated from human donors that have been diagnosed with diabetes type I or type II disease. Diseased cells are grown in the same optimized media system as normal human endothelial cells.
Diabetic cultures can be used for experimental applications in diabetic-related cardiovascular pharmaceutical development, vascular pathology, including atherosclerosis.
Cryopreserved Clonetics® Diabetic human coronary artery endothelial cells are shipped in third passage. Proliferating Clonetics® cells are shipped in fourth passage. All cells test negative for mycoplasma, bacteria, yeast, and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots.
Delivery: Content: 1×cryopreserved Ampule with ≥500000 cells/vial and Certificate of Analysis is provided for each cell lot purchased.
Customers Who Bought This Also Bought