D-HAEC – Diseased Human Aortic Endothelial Cells – Diabetes Type I
Catalog #: CA10064-318
Supplier: LONZA PHARMA - BIOSCIENCE CA
CAS Number:
Specifications
- Description:D-HAEC – Diseased Human Aortic Endothelial Cells – diabetes type I
- Size:≥500000 cells
- Cat. no.:CA10064-318
Specifications
About this item
Diseased human aortic endothelial cells are isolated from human donors that have been diagnosed with diabetes type I disease. Diseased cells are grown in the same optimized media system as Normal Human Endothelial Cells.
Diabetic cultures can be used for experimental applications in diabetic-related cardiovascular pharmaceutical development, vascular pathology, including atherosclerosis. Cryopreserved Clonetics® Diabetic Human Aortic Endothelial Cells are shipped in third passage. Proliferating Clonetics® cells are shipped in fourth passage.
All cells test negative for mycoplasma, bacteria, yeast, and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots.
Delivery: Content: 1×cryopreserved Ampule with ≥500000 cells/vial and Certificate of Analysis is provided for each cell lot purchased.
Customers Who Bought This Also Bought