NHEK-Ad - Human Epidermal Keratinocytes, Adult, Single Donor
CA10063-800
:
- Description:NHEK-Ad - Human Epidermal Keratinocytes, Adult, single donor
- Size:≥500000 cells
- Environmentally Preferable:
- Cat. no.:CA10063-800
Cryopreserved normal human epidermal keratinocytes are guaranteed through 18 population doublings for adult cells.
All cells test negative for mycoplasma, bacteria, yeast, and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots. Normal Human Dermal fibroblasts from matched donors are available for certain lots.
: Content: 1×Cryopreserved ampule of NHEK-Ad from single donor containing ≥500000 cells and Certificate of Analysis is provided for each cell lot purchased.