ContactUsLinkComponent
Human Mesenchymal Stem Cell (hMSC) Chondrogenic Differentiation Medium BulletKit®
Human Mesenchymal Stem Cell (hMSC) Chondrogenic Differentiation Medium BulletKit®
Catalog #: CA10064-090
CAS Number:  
Kit hMSC - Human Mesenchymal Stem Cell Chondrogenic Differentiation Medium
Human Mesenchymal Stem Cell (hMSC) Chondrogenic Differentiation Medium BulletKit®
Catalog #: CA10064-090
CAS Number:  
Specifications
  • Description:
    Human Mesenchymal Stem cell (hMSC) chondrogenic differentiation medium Bulletkit®
  • Cat. no.:
    CA10064-090
Specifications
About this item
Stem cells possess the unique ability to differentiate into multiple lineages of cells including chondrocytes.
  • Directly visualize bands for recovery
  • See bands in as little as 2 minutes
  • Recover samples directly without UV light band excision or purification
  • Real-time separation and documentation
  • Watch band migration as it happens
  • Photograph gels at the bench without DNA damaging UV light
  • Outstanding sensitivity and resolution
Lonza’s hMSC chondrogenic differentiation BulletKit® Medium is a serum containing medium designed to induce chondrogenic differentiation, with the addition of TGF-β3, of human bone marrow derived mesenchymal stem cells into mature, functionally active chondrocytes in as few as 14 days. Differentiation can then be validated by a variety of methods including staining with alcian blue or safranin.
Partial list of cells differentiated with hMSC chondrogenic differentiation BulletKit® medium*:
Human bone marrow derived stem cells.
Human umbilical cord blood derived stem cells.
Human umbilical cord matrix derived stem cells.
C57BL/6 mice bone marrow derived stem cells.
human adipose derived stem cells.
Human hamstring tendon derived stem cells.
The hMSC Chondrogenic Differentiation BulletKit® medium, with the addition of TGF-β3 (sold separately) provides a differentiation medium guaranteed to induce chondrogenic differentiation of bone marrow derived hMSCs into mature, functionally active chondrocytes.Each bottle of the hMSC Chondrogenic Differentiation BulletKit® medium contains enough differentiation medium to fully differentiate at least 2 million bone marrow derived hMSC [approximately 4 complete 24-well plates] into mature, functionally active chondrocytes.
The hMSC Chondrogenic differentiation medium is offered as a BulletKit® medium (catalog no. PT-3003) which includes both the basal media and the necessary supplements (with the addition of TGF-β3 [sold separately, catalog no. PT-4124]) for chondrogenic differentiation of bone marrow derived hMSCs. The basal medium (Differentiation Basal Medium - Chondrogenic; catalog no. PT-3925) and the necessary supplements (HMSC SQ Kit - Chondro; catalog no. PT-4121 and TGF Beta 3 for HMSC Chondro diff; catalog no. PT-4124) can also be purchased individually.
Lonza guarantees the performance of cells only if appropriate media and reagents are used exclusively and the recommended storage and use protocols are followed. Any modifications made to the recommended cell systems including the use of alternative media, reagents, or protocols, will void cell and media performance guarantees.
Delivery: Content:
1×hMSC Chondrogenic Basal Medium (PT-3925).
1×hMSC Chondrogenic SingleQuot kit (PT-4121).
Customers Who Bought This Also Bought