About this item
The ToxiLight® 100% lysis reagent set is intended for use with the ToxiLight® non-destructive cytotoxicity assay (sold seperately) and is designed to give the user a 100% lysis or total adenylate kinase control.
- Highly sensitive - The ToxiLight® BioAssay Kit exploits the cyclic nature of the AK reaction whereby a small amount of the AK enzyme can generate high concentrations of ATP. This enables the detection of very subtle changes in cytotoxicity and reduces false negatives
- Non-destructive - ToxiLight® assay detects cellular AK present in cell culture supernatants, eliminating the need to lyse cells to perform the assay, and allowing multiple tests to be performed on the same sample
- Simple - Assay requires the simple addition of a single reagent, which can be added directly to wells in which cells are growing, or to a small sample of aspirated culture supernatant
- Fast - Readable signal is detectable 5 minutes after the addition of the ToxiLight® BioAssay reagent
- Flexible - Cell supernatants can be frozen with no loss of AK activity, allowing for long-term kinetic studies
- Safe - No hazardous chemicals are required
It contains the 100% Lysis Reagent, 10 ml and tris-acetate buffer, 50 ml (required volume correction for sample wells), and is suitable for 200 tests in a 96 well format.
Simply add the 100% lysis reagent to the control wells at a ratio of 1:2 (lysis reagent : sample volume), volume correct the remaining sample wells with Tris-Acetate Buffer and then incubate at room temperature for 10 minutes.
The ToxiLight® 100% lysis reagent should be included as a control at each time point within your cytotoxicity assay. After addition proceed with the standard ToxiLight® assay protocol.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Related Information
ToxiLight™ 100% Lysis Reagent Set
Application Notes
ToxiLight™ Bioassay Kit Non Destructive Cytotoxicity Assay