About this item
The ToxiLight® Bioassay kit is a bioluminescent, non-destructive cytolysis assay kit designed to measure the release of the enzyme, adenylate kinase (AK), from damaged cells. AK is a robust protein present in all eukaryotic cells, which is released into the culture medium when cells die.
- Highly sensitive - The ToxiLight® BioAssay kit exploits the cyclic nature of the AK reaction whereby a small amount of the AK enzyme can generate high concentrations of ATP. This enables the detection of very subtle changes in cytotoxicity and reduces false negatives
- Non-destructive - ToxiLight® assay detects cellular AK present in cell culture supernatants, eliminating the need to lyse cells to perform the assay, and allowing multiple tests to be performed on the same sample
- Simple - Assay requires the simple addition of a single reagent, which can be added directly to wells in which cells are growing, or to a small sample of aspirated culture supernatant
- Fast - Readable signal is detectable 5 minutes after the addition of the ToxiLight® BioAssay reagent
- Flexible - Cell supernatants can be frozen with no loss of AK activity, allowing for long-term kinetic studies
- Safe - No hazardous chemicals are required
The enzyme actively phosphorylates ADP to form ATP and the resultant ATP is then measured using the bioluminescent firefly luciferase reaction. As the level of cytolysis increases, the amount of AK in the supernatant also increases, which results in emission of higher light intensity by the ToxiLight® reagent. Because the ToxiLight® Bioassay Kit exploits the fact that AK is released from cells when they die, there is no need for cell lysis (unlike many other cytotoxicity assays). Repeated samples of supernatant can therefore be taken over time without disrupting the cells themselves. This allows for kinetic analysis of cell death.
The ToxiLight® BioAssay Kit also facilitates high content screening by allowing other tests to be performed on the original cells.
All the components required for AK detection are contained within a single reagent making the assay very simple to perform. The kit provides outstanding sensitivity with a detection limit of 10 cells per microwell with a dynamic range of over 5 orders of magnitude. Extended signal stability also makes the assay suitable for batch processing in high-throughput screening applications.
The ToxiLight® 100% Lysis Reagent set is intended for use with the ToxiLight® Non-destructive Cytotoxicity Assay and is designed to give the user a 100% lysis or total adenylate kinase control. It contains the 100% Lysis Reagent, 10 ml and Tris-Acetate Buffer, 50 ml (required volume correction for sample wells), and is suitable for 200 tests in a 96 well format. ToxiLight® 100% Lysis Reagent Set, LT07-517. (Available Separately).
Simply add the 100% Lysis Reagent to the control wells at a ratio of 1:2 (lysis reagent : sample volume), volume correct the remaining sample wells with Tris-Acetate Buffer and then incubate at room temperature for 10 minutes. The ToxiLight® 100% Lysis Reagent should be included as a control at each time point within your cytotoxicity assay. After addition proceed with the standard ToxiLight® Assay protocol.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Related Information
ToxiLight™ 100% Lysis Reagent Set
Application Notes
ToxiLight™ Bioassay Kit Non Destructive Cytotoxicity Assay