To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Latitude® midigel, 2%
- Size:8 gels
- Cat. no.:CA11007-622
- Supplier No.:57231
Specifications
About this item
Precast Latitude® midigels TBE are made for accurate DNA fragment separation for most any gel electrophoresis application. They are manufactured from high quality, Lonza molecular biology grade SeaKem® LE Plus Agarose.
- Fast - No more pouring or cooling times, easy - simply unwrap and use, traceable - trackable lots
These ready-to-run gels are available in two different well formats and agarose concentrations in TBE buffer to fit most DNA electrophoresis needs. Our Latitude® Precast Agarose Gels are precision cast for accurate, reproducible resolution of DNA fragments for 100 bp to 8 kb and pre stained with 0.5 µg/ml Ethidium Bromide. Latitude® Midigels TBE fit most standard electrophoresis chambers and are stable for 6 to 12 months, depending on their concentration.
Specifications:
Separation range 100 bp to 2 kb
Gel dimensions 10×15 cm
Gel thickness 6.0 mm
Ethidium bromide 0.5 μg/ml
Tray dimensions 10.4×15.6 cm
Well volume10 to 12 μl
Applications: DNA Electrophoresis.