VWR® Blue Light Nitrile Examination Gloves
Supplier: VWR International
Certificates
About this item
Blue light nitrile examination gloves offer a thin 2 mil finger thickness that provides protection against a broad range of chemicals. These gloves are made of premium compounded nitrile for high manufacturing quality measures and product consistency.
- 2 mil finger thickness
- Powder-free
- 9.5" length
- Food contact compliant
- High comfort and flexibility
- Finger textured
Blue light nitrile examination gloves are 100% latex-free to eliminate adverse reactions associated with natural rubber latex proteins. The gloves have a beaded cuff that allows for easy donning and a rolled finger texture for added grip. They are ideal for use in industrial and laboratory applications where protection against chemicals and biohazards is required. The gloves meet or exceed ASTM D6319 standards for water leak and dimension.
Packaging: Gloves are packaged 100 gloves per box, 10 boxes per case.
Specifications
- Catalog No:
- 76717-018
- 76717-020
- 76717-012
- 76717-014
- 76717-016
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Material:
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- EN norm:
- EN 455 certified
- EN 455 certified
- EN 455 certified
- EN 455 certified
- EN 455 certified
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Food contact approved:
- Yes
- Yes
- Yes
- Yes
- Yes
Specifications
Customers Who Bought This Also Bought