Saint Gobain® Sanitary Tri-Clamps to Hose Barb Adapter
Supplier: Saint Gobain Life Sciences
New Product
Certificates
About this item
Quick and secure sanitary tubing attachments are critical in numerous low-pressure laboratory, biotechnology, and pharmaceutical applications. Contamination of fluids and product loss through leakage can be costly. These Sanitary Fittings provide a superior connection to tubing through a single-barb construction, reducing the potential for leakage.
- Tapered funnel design reduces 'dead space' for exceptionally smooth fluid flow
- Smooth inner surface prevents particle entrapment
- Seamless construction prevents contamination by eliminating leaks and areas of potential bacterial growth
- Purchase alongside flexible tubing, ½" sanitary gasket, and a mini tri-clamp to complete the assembly
Fittings are available in Kynar (PVDF), polypropylene (PP), and polysulfone (PS). These materials are all radiation sterilizable. Sterilize by gamma irradiation. Produced from materials that meet USP Class VI.
Customers Who Bought This Also Bought