Specifications
- Capacity:4.7 ml
- Max. pressure:3 bar [0.3 MPa] (44 psi)
- Description:HiScreen™ MabSelect™ VL
- Binding capacity:~70 mg human IgG/ml resin at 6 min residence time
~60 mg human IgG/ml resin at 4 min residence time - Storage:2...8 °C, 20% Ethanol
- Bed Dimensions:7.7×100 mm
- Flow Velocity:<300 cm/h
- Cat. no.:CA76716-902
Specifications
About this item
MabSelect™ VL protein L resin in HiScreen™ chromatography columns. A redesigned protein L ligand captures antibody Fabs, bispecifics, and other variants containing a kappa light chain with high binding capacity and improved alkaline stability.
- High dynamic binding capacity for efficient capture of target molecules
- Stable when cleaned with 0.1 M NaOH, reducing risk for bioburden incidents
- Provides good resolution for product-related impurities in the capture of bispecific antibodies
- MabSelect™ VL protein L resin showed a two-fold increase in binding capacity compared to our Capto™ L chromatography resin
- This higher capacity provides productivity advantages in research and biomanufacturing
- The MabSelect™ VL protein L resin can withstand cleaning with 0.1 M NaOH
As the diversity of antibody variants increases, so does the need for chromatography resins that can capture and purify these variants. As an alternative to protein A, protein L resin can be used to capture antibody Fabs, bispecific antibodies, and other molecules containing kappa variants of the variable light (VL) chain. Protein L binds human kappa light chain subtypes 1, 3, and 4 and mouse kappa light chain subtype 1.
HiScreen™ columns are convenient for method optimization and parameter screening enabled by the 10 cm bed height for efficient chromatography process development. MabSelect™ VL protein L resin is also available in 1 and 5 ml HiTrap™ prepacked columns, bulk resins in different pack sizes, PreDictor™ RoboColumn® minicolumns, and ReadyToProcess™ columns for purifying protein as well as screening and process evaluation.