About this item
Cage washing is a critical step in ensuring the safest possible biomedical environment for your staff, animals, and valuable research. Explore our portfolio of professional enzyme, alkaline, and acid cage wash detergent solutions for your life sciences facility.
- Versatile, low-foaming detergent specifically designed for the removal of inorganic residues and salts
- Highly effective at the removal of scale, hard water deposits, animal fats, and dried feces on animal cages and other equipment
- Designed for manual and automated cage washers. Can be used in both mechanical and soak applications
- Stabilized and broad dilution ratio-based formula to enhance cleaning and reduce rusting and corrosion
- Used in conjunction with Alka Gent® alkaline cage wash detergent to equalize residual pH and provide excellent rinse-through results
- Buffer system provides two modes of cleaning power
- Detergent polymer stops bio-burden from depositing on instruments
For use in both automated cage washers, manual use on a variety of equipment and ultrasonic units within the life science, veterinary, and research industries.