Knife Switch, Small
Catalog #: 470353-956
Supplier: United Scientific Supplies
New Product
Specifications
- Description:Knife switch, screw posts, single pole, single throw
- Cat. no.:470353-956
Specifications
About this item
Tradiitonal lever switch for circuit experiments.
- Easy pivot lever
- Secure connection screws
Single-pole, single-throw knife switch with plastic base is suitable for low voltage experiments (less than 3.6 V). Includes thumbscrew connecting posts.
Customers Who Bought This Also Bought