About this item
General-purpose syringes available in 1, 3, 5, 10 and 20 ml barrel volumes for clinical and veterinary use.
- EtO sterilized
- Pyrogen-free
- Highly transparent barrel for maximum visibility
- Excellent lubrication performance
- Plunger retention mechanism prevents accidental plunger withdrawal
These syringes are non-reactive and safe for use in medical procedures. The barrel and plunger are made from polypropylene, and the piston is made from polyisoprene. All syringes do not contain any DEHP, BPA, or styrene. This makes them suitable for clinical use including injecting fluid into the body and collecting fluid from the body.