Staticide® 100NI Blue Nitrile ESD Finger Cots
Supplier: ACL Staticide
New Product
Certificates
About this item
Staticide blue nitrile ESD finger cots provides superior static protection and chemical resistance without latex.
- Powder-free
- Suggested for Class II static sensitivity devices
- ISO 5/Class 100 cleanroom compatible
- 3 mil thickness
Nitrile finger cots provide inherent static resistance. 10⁹ ohms resistance with CAFÉ testing.
Packaging: Bags of 720 cots are vacuum sealed. 4 bags per case.
Customers Who Bought This Also Bought