Plastic Pails with Plastic Handle, UN Rated, 14.2 L (3.76 Gallon)
Supplier: Starplex
Certificates
About this item
Pail constructed of polypropylene and lid is made of high-density polyethylene.
- UN Rated
- Available in black or white
Customers Who Bought This Also Bought