Pipet Keeper™ XL Biohazard Waste Container
Catalog #: 77321-812
Supplier: Whitney Products
New Product
Specifications
- Capacity:4200 cm³
- Dimensions:76×114×419 mm (3×4.5×16.5")
- Description:Biohazard Pipet Keeper™ XL container
- Material:Sturdy paperboard and plastic-lined interior
Specifications
About this item
Pipet Keeper XL biohazard collection containers are designed to conveniently collect up to 16" long biohazardous waste at your workstation.
- Dispose of pipettes, swabs and other long, biohazardous objects easily
- Sturdy construction - made of sturdy paperboard printed with orange biohazard symbols
- Plastic-lined interior holds a moderate amount of liquid
- Self-locking tops keep contents safely enclosed
- Flat storage requires far less space than rigid plastic containers
- Interlocking flaps - easily fold into shape
Collects waste up to 16" - allows quick and efficient disposal without the need for additional handling or transportation.
Customers Who Bought This Also Bought