Eisco Glass Separatory Funnels with Screw-Type Stopcock
Supplier: Wards
Certificates
About this item
Separatory funnels with ground cones, plastic stoppers, and Rotaflow screw-type stopcocks, made of borosilicate 3.3 glass.
- Made of high quality, heavy-duty borosilicate 3.3 glass - can withstand high temperatures and thermal shocks
- Used to separate the components of a mixture into two immiscible liquids of different densities
- Fitted with a screw-type Rotaflow stopcock for precise drop-wise dispensing of the denser separated liquid
- Features a ground glass cone with and interchangeable plastic stopper
- Ungraduated
Ungraduated, pear-shaped Separating Funnels are made of high quality borosilicate glass. They feature a ground cone, plastic stopper, and screw-type Rotaflow stopcock. Autoclavable, with superior chemical durability. Low temperature gradient - can withstand high temperatures and thermal shocks.
Eisco Labs borosilicate glass has the following heat tolerances:
- Strain Point 515 °C
- Annealing Point 565 °C
- Softening Point 820 °C
Customers Who Bought This Also Bought