Dispensette® S Bottle Top Dispensers
Supplier: Brandtech
Certificates
About this item
Fixed volume or variable volume, analogue-adjustable or digital easy calibration.
- With or without recirculation valve
- Autoclavable at 121 °C
- Three different models of volume adjustment available: Fixed (fixed volume for standard applications; simple calibration adjustment with supplied tool) and Analogue (analogue volume display, fast volume adjustment; simple calibration adjustment with supplied tool)
- Digital EASY CALIBRATION: Easy to read digital display; dispensing volume can be set accurately and reproducibly. Easy calibration technique for adjustment in seconds without tools; alteration of factory setting is automatically indicated
- Easy to calibrate and adjust in order to comply with ISO 9001 and GLP guidelines; alteration of factory setting is automatically indicated
Dispensette® S supports a wide range of applications for the dispensing of aggressive reagents directly from the supply bottle; such as concentrated bases and acids like H₃PO₄, H₂SO₄ (with certain exceptions such as HCl, HNO₃, HF, and so on), saline solutions, and a variety of organic solvents. Parts that are in contact with the liquid are made from borosilicate glass, ceramic, platinum-iridium, ETFE, FEP, PFA, PTFE and PP (discharge tube safety screw cap). Colour code: Red.
* Special fixed volumes: 0.5 to 100 ml (please state when ordering)!
Ordering information: Dispensette® S is supplied with a DE-M marking, performance certificate, telescopic filling tube, recirculation tube (optional), mounting tool and PP adapters.
Specifications
- Catalog No:
- 10019-016
- 10018-978
- 10020-960
- 10018-936
- 10018-980
- 10018-976
- 10018-942
- 10018-944
- 10020-966
- 10018-938
- 10020-964
- 10018-940
- 10020-962
- 10018-930
- 10018-972
- 10018-932
- 10018-974
- 10018-968
- 10019-012
- 10018-970
- 10019-014
- 10018-934
- 10018-956
- 10018-958
- 10018-964
- 10018-966
- 10019-010
- 10018-960
- 10018-962
- 10018-950
- 10018-952
- 10018-946
- 10018-948
- 10018-954
- Environmentally Preferable:
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
Specifications
Customers Who Bought This Also Bought