Eisco Plastic Filter Funnels
Supplier: Wards
Certificates
About this item
Chemically-resistant filter funnels made of high quality polyethylene plastic.
- Excellent resistance against acids, alcohols, aldehydes, bases, and hydrocarbons
- Useful for separating solids from liquids, as well as for easier transfers of liquids to narrow mouthed containers
- Autoclavable and microwavable
Packaging: Pack of 1 or 10.
Customers Who Bought This Also Bought