Sapphire Single and Multichannel Pipettes
Supplier: Greiner Bio-One
Certificates
About this item
Sapphire pipettes are a variable volume range of air-displacement pipettes designed and built to deliver optimal performance, ergonomics, and robustness in day-to-day use.
- Unique serial number
- Fully autoclaveable
- Available in single and multichannel models with a volume range from 0.2 µl to 10 ml
- Ergonomic design for optimum comfort
The lightweight design coupled with a state-of-the-art mechanism reduces pipetting forces and makes our pipettes both comfortable and easy-to-use. The contoured body shape allows for a relaxed grip whatever size your hand, and the 3-position tip ejector button makes pipetting comfortable for both left-handed and right-handed users.
Customers Who Bought This Also Bought