Oakton® pH 150 Waterproof Portable pH/mV/Temperature Meter
Catalog #: 76221-264
Supplier: Argos Technologies
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Calibration:Up to 6 points
- Description:Waterproof portable meter kit
- Temperature range:−10...+110 °C
- IP rating:IP 67
- Automatic buffer recognition:DIN, NIST, and USA
- Connections:BNC
- Dimensions:79.4 × 57.2 mm (3¹/₈ × 2¹/₄")
- Display:3 line custom LCD
- mV accuracy:±0.2
- mV range:−2000...+2000 mV
- mV resolution:0.1 mV for ±999.9 mV; 1 mV beyond ±1000 mV
- pH accuracy:±0.01
- pH range:−2...+16
- pH resolution:0.01
- Cat. no.:76221-264
Specifications
About this item
Rugged, accurate, reliable - Best value for an IP 67-rated waterproof meter.
- Intuitive, user-friendly icons - electrode status displays pH slope
- Grip-Clip™ holder quickly and easily secures the electrode to your beaker or container
- Expanded data logging for up to 150 data sets
- mV offset for calibration of ORP standards, zeroing mV value, and pH diagnostics
- 700 hours battery-life or optional universal power adapter
Built-in stand and wall mount option allow for easy benchtop measurement or storage.
Delivery: Meter includes Grip-Clip holder and batteries; meter with probe adds on electrode 76221-306; kit further adds buffers, solutions, and hard carrying case.
Customers Who Bought This Also Bought