TaskBrand® V40 Value Series DRC Wipes
Supplier: High-Tech Conversions
New Product
Certificates
About this item
DRC is the leading all-purpose disposable wipe.
- All-purpose disposable wipe
- Highly absorbent dry wipe
- Heavy-duty, low linting
- Unique stretch with cloth-like feel
- Made of 35% (avg.) post-industrial recycled fibers
TaskBrand® V40 value series DRC wipes are made of 35% post-industrial recycled fibers, making up a minimum 85% of the total product weight. Applications include glass and mirror cleaning, equipment/tools/parts cleaning, staining, wiping up oil/grease/water, general-purpose surface and hand cleaning, janitorial and facility maintenance applications.
Customers Who Bought This Also Bought