Virtu-Clean® Disposable Cleaning Pads with Scrubber
Catalog #: 77008-034
Supplier: High-Tech Conversions
New Product
Specifications
- Description:Virtu-Clean® disposable cleaning pad with scrubber
- Cat. no.:77008-034
- Packaging:20 pads/bag, 12 bags/master bag
Specifications
About this item
Not all cleaning pads are equal. Absorbency rates, release rates, coverage, and linting can vary greatly, significantly impacting the finished product.
- Cleanroom class: ISO 5 (Class 100)
- Scrubber pad option is designed for more aggressive cleaning and stain removal
- Holds 7X its weight in cleaning fluid
- Charge-once-and-clean coverage area of 240 to 300 sq. ft.
- No Quat binding
- Compatible with chlorine bleach, hydrogen peroxide, and common disinfectants
Virtu-Clean™ Disposable Cleaning Pads are specially engineered for critical environments, providing maximum coverage, cleaning, and convenience. These disposable, single-use mop pads check all the right boxes – they hold more liquid, release that liquid at a controlled rate over a larger surface area, and reduce friction for more effort-free cleaning.
Customers Who Bought This Also Bought