ProWorks® Catch® Black Nitrile Powder-Free Gloves
Supplier: High-Tech Conversions
New Product
Certificates
About this item
ProWorks® Catch® Black nitrile powder-free disposable gloves feature a heavy pyramid Grip® textured surface that provides an impeccable grip.
- Industrial grade nitrile construction
- Powder-free and latex-free
- 9 mil thickness
- Ambidextrous
The extra thickness makes these gloves more durable when working on 'rougher' jobs, while keeping hands safe and protected against harsh chemicals, solvents, and paints.
Certifications: Complies with FDA CFR Title 21 Indirect Food Additive Regulations Part 177.
Packaging: Gloves are packaged 100 pieces per box.
Specifications
- Catalog No:
- 77007-974
- 77008-024
- 77007-874
- 77007-970
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Material:
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile