VWR® Media Bottles, Square, PETG
Supplier: VWR International
Certificates
About this item
Square PETG bottles are ideal for storage of media, laboratory reagents and biological buffers up to −70 °C. Eliminate washing, depyrogenation and autoclaving steps with sterile and non-pyrogenic.
- Made from 100% virgin PETG material, durable and break resistant
- Heat-shrink band around closure and neck for tamper-evident seal
- Leakproof bottles and HDPE closures provided
- Leakproof sealing with integrated lip seal feature
- Sterile and non-pyrogenic, SAL 10⁻⁶
- Low extractables
- Transparency and reduced permeability
- Arranged in shrink-wrapped trays and polybag for prevent contamination
- 2 L product come with in mold hand grip for better ergonomics
- Products 30 ml and above has volume graduation
Certifications: US FDA 21CRF, USP CLASS VI compliant Resin.
BodyTextAccessories: Spare closures are available as per specific application requirements.
Packaging: Bottles are packed with prefer band, tray arranged and poly bagged. Spare Closures are bulk packed.
Related Information
Certificates
Search by Lot NumberFrozen Serum Cell Growth Test Hydrogen Peroxide Permeability Test PETG Bottles Extractables and Leachable
Application Notes
Regulatory Compliance VWR PETG Bottles
Conformity declaration
Customers Who Bought This Also Bought