VWR® Square Polycarbonate Media Bottles
Supplier: VWR International
Certificates
About this item
VWR® square polycarbonate bottles are ideal for storage of media, laboratory reagents, and biological buffers in temperatures ranging from −130 to +121 °C. Eliminate washing, depyrogenation, and autoclaving steps with sterile and non-pyrogenic bottles.
- Made from durable 100% virgin polycarbonate material
- Suitable for frozen storage down to −130 °C
- Heat-shrink band around closure and neck for tamper-evident seal
- Leakproof bottles and closures (HDPE/PP) provided for applicable products
- Leakproof sealing with integrated lip seal feature
- Sterile and non-pyrogenic, SAL 10⁻⁶
- Low extractables
- Transparency and reduced permeability
Bottles are arranged in shrink-wrapped trays and polybag to prevent contamination. 2 L bottles feature a molded hand grip for better ergonomics. Bottles 30 ml and above have volume graduations.
Certifications: US FDA 21CRF, USP CLASS VI compliant Resin.
BodyTextAccessories: Spare closures are available for specific application requirements. See part numbers VWRI210-000111, VWRI210-000112, VWRI210-000113, VWRI210-000114, VWRI210-000115, VWRI210-000116, VWRI210-000117, VWRI210-000118, VWRI210-000119 and VWRI210-000120.
Packaging: Bottles are packed with band, tray arranged, and poly bagged. Spare closures are bulk packed.
Customers Who Bought This Also Bought