Protect 800™ Sterile Nitrile Chemo Examination Gloves
Supplier: High-Tech Conversions
New Product
Certificates
About this item
Sterile chemo gloves are the first line of defense against the harmful effects of HDs.
- Textured fingertips
- Extended cuffs
- Latex-free
The Protect 800™ chemo gloves are ambidextrous, powder-free, and made of nitrile. These gloves are perfect for cancer centers, administration of chemo, handling of HDs, and for compounding chemo. The gloves are packaged in 100% poly bags for lint-free donning. The fingertips are textured for extra grip, and the gloves feature extended cuffs for added protection. These gloves have been tested for a range of chemotherapy drugs.
Packaging: Gloves are packaged 50 pair/bag, 4 bags/pack, 200/case.
Specifications
- Catalog No:
- 77007-924
- 77007-838
- 77007-878
- 77007-980
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Material:
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Sterility:
- Sterile
- Sterile
- Sterile
- Sterile