Precision® Blue Nitrile Powder-Free Examination Gloves
Supplier: High-Tech Conversions
New Product
Certificates
About this item
Ultra-soft and super stretchy Precision® Blue Nitrile Powder-Free Exam Gloves feature a unique low modulus formulation that reduces hand fatigue and provides maximum comfort.
- 100% synthetic and latex-free
- Thinner design for maximum dexterity
- Strong formulation provides superior puncture and tear resistance
- Excellent barrier protection
Gloves are 100% synthetic and latex-free, avoiding potential allergic reactions to latex proteins. Precision® Nitrile Gloves are made thinner for maximum dexterity, yet they maintain strength to provide superior puncture and tear resistance. These gloves offer excellent barrier protection.
Packaging: Gloves are packaged 100 pieces per box (90 pieces per box for size XL).
Specifications
- Catalog No:
- 77007-886
- 77007-930
- 77007-844
- 77007-832
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Material:
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile