About this item
Printer optimized for 1-, 2- or 4-color panel ribbons for multicolor made easy - no calibrating, no threading, no waste (single-color ribbon only in kit). The S3700 empowers any user to create any way they choose with time-saving onboard label templates, intuitive PC labeling software.
- Cutter type: XY cutter and auto cutter
- Multi-color printing capability
- 600 labels per day (max.)
Create shapes, arrows, letters and more with the built-in XY plotter cutter that slices straight lines, smoother curves and tighter connection points. Get your print jobs done right in less time with 20-second ribbon and material changeovers plus up to 5-in.-per-second print speed. Empower any user to print how they choose with time-saving onboard templates, intuitive PC labeling software and a mobile app.
Color kit - Get all the colors and capabilities you need in one convenient kit to create safety labels, lean visuals, lockout tags, work-in-process labels and more that truly pop.
Starter kit - Create professional, impactful safety tags by adding your specific hazard information to these print-on, write-on tags. Durable, low-shrink all weather permanent adhesive vinyl label tape with ultra-aggressive adhesive is ideal for just about any industrial surface.
Lean kit - Repositionable magnetic labels and printable ToughStripe® floor tape are perfect for bin labeling, area marking and other lean/5S applications. Durable, low-shrink all weather permanent adhesive vinyl label tape with ultra-aggressive adhesive is ideal for just about any industrial surface.
GHS kit - Tough, smear-proof ribbon produces long-lasting printed text that can withstand years in industrial areas and outdoor conditions. Durable pre-printed GHS labels contain ultra-aggressive adhesive to withstand exposure to moisture, chemicals and handling.
ARC kit - Durable, low-shrink all weather permanent adhesive vinyl labels with ANSI WARNING header will last and help keep you compliant. Raised panel labels are designed for patch panel identification, external push-buttons, switches and internal connection points.
Pipe kit - Simply tap, print, go with preprogrammed onboard label templates plus 600 barcodes and symbols. Durable, low-shrink all weather permanent adhesive vinyl label tape with ultra-aggressive adhesive is ideal for labeling pipes indoors and out.
M611 kit – Desktop and portable printer in one kit. The M611 lets you create, share and print up to 2" width labels at 2" per second print speed anywhere, anytime.
Printer starter kit includes (1) S3700 Safety and facility printer (S3700-WB);(1) Brady workstation safety and facility ID software (BWS-SFIDS-NA-EM);(1) Cleaning kit (PCK-6);(1) Mouse;(1) Power cord;(1) Quick start guide;(1) USB cord;(1) USB flash drive;(1) 4" Continuous white vinyl label (B30C-4000-595-WT);(1) 2.25" Continuous white vinyl label (B30C-2250-595-WT);(1) 1.125" Continuous white vinyl label (B30C-1125-595-WT);(1) 4" Continuous yellow vinyl label(B30C-4000-595-YL);(1) 2.25" Continuous yellow vinyl label (B30C-2250-595-YL;(1) 4×6" White vinyl label (B30-25-595-BLNKWT);(1) OSHA DANGER tags (B30-255-851-WHT);(2) Black ribbon cartridges (B30-R10000);(1) White ribbon cartridge (B30-R10000-WT);(1) Red ribbon cartridge (B30-R10000-RD).
Lean/5S label kit includes (1) S3700 Safety and facility printer (S3700-WB);(1) Brady workstation safety and facility ID software (BWS-SFIDS-NA-EM);(1) Cleaning kit (PCK-6);(1) Mouse;(1) Power cord;(1) Quick start guide;(1) USB flash drive;(1) 4" White vinyl continuous label (B30C-4000-595-WT);(1) 2.25" Continuous white vinyl label (B30C-2250-595-WT);(1) 4" Continuous yellow label (B30C-4000-595-YL);(1) 2.25" Continuous yellow vinyl label (B30C-2250-595-YL);(1) 3.85×2.25" White vinyl label (B30-219-595-BLNKWT);(1) 2.5" Coninuous magnetic polyester label (B30C-2500-509-WT);(1) 4.25" Continuous magnetic polyester label (B30C-4250-509-WT);(1) 2.25" Continous yellow polyester label (B30C-2250-483YL-KT);(1) Black ribbon cartridge (B30-R10000);(1) White ribbon cartridge (B30-R10000-WT);(1) Red ribbon cartridge (B30-R10000-RD);(1) Blue ribbon cartridge (B30-R10000-BL);(1) 4-color ribbon cartridge (B30-R10000-KRYG-8).
GHS labeling kit includes (1) S3700 Safety and facility printer (S3700-WB);(1) Brady workstation safety and facility ID software (BWS-SFIDS-NA-EM);(1) Cleaning kit (PCK-6);(1) Mouse;(1) Power cord;(1) Quick start guide;(1) USB cord;(1) USB flash drive;(1) Pre-printed GHS labels (B30-262-7569-CLP4B);(1) Pre-printed GHS labels (B30-260-7569-CLP4B);(2) Black ribbon cartridges (B30-R10000).
Arc flash labeling kit includes (1) S3700 Safety and facility printer (S3700-WB);(1) Brady workstation safety and facility ID software (BWS-SFIDS-NA-EM);(1) Cleaning kit (PCK-6);(1) Mouse;(1) Power cord;(1) Quick start guide;(1) USB flash drive;(1) ANSI WARNING vinyl labels (B30-25-595-ANSIWA);(1) Raised panel polyester labels (B30EP-174-593-WT);(1) 2.25" Continuous orange vinyl label (B30C-2250-595-OR);(2) Black ribbon cartridges (B30-R10000).
Pipe marking kit includes (1) S3700 Safety and facility printer (S3700-WB);(1) Brady workstation safety and facility ID software (BWS-SFIDS-NA-EM);(1) Cleaning kit (PCK-6);(1) Mouse;(1) Power cord;(1) Quick start guide;(1) USB cord;(1) USB flash drive;(1) 4" Continous yellow vinyl labels (B30C-4000-595-YL);(1) 2.25" Continous vinyl label (B30C-2250-595-YL);(1) 4" Continous green vinyl labels (B30C-4000-595-GN);(1) 2.25" Continous green vinyl labels (B30C-2250-595-GN);(1) 4" Continous red vinyl (B30C-4000-595-RD);(1) 2.25" Continous red vinyl labels (B30C-2250-595-RD);(1) 4" Continous blue vinyl labels (B30C-4000-595-BL);(1) 2.25" Continuous blue vinyl labels (B30C-2250-595-BL);(1) 4" Continous white vinyl labels (B30C-4000-595-WT);(1) Black ribbon cartridge (B30-R10000);(3) White ribbon cartridges (B30-R10000-WT).
Desktop and portable printer includes (1) S3700 Safety and facility printer (S3700-WB);(1) Brady workstation safety and facility ID software (BWS-SFIDS-NA-EM);(1) Cleaning kit (PCK-6);(1) Mouse;(1) Power cord;(1) Quick start guide;(1) USB cord;(1) USB flash drive;(1) 4" Continuous white vinyl labels (B30C-4000-595-WT);(1) 4" Continous yellow vinyl labels (B30C-4000-595-YL);(1) 2.25" Continous white vinyl labels (B30C-2250-595-WT);(1) 2.25" Continous yellow vinyl labels (B30C-2250-595-YL);(2) Black ribbon cartridges (B30-R10000);(1) 1" Continous white vinyl labels (M6C-1000-595-WT);(1) 1" Continuous yellow vinyl labels (M6C-1000-595-YL);(1) 0.5" Continuous white vinyl labels (M6C-500-595-WT);(1) 0.5" Continuous yellow vinyl labels (M6C-500-595-YL);(2) Black ribbon cartirdges (M61-R10010); M611 Bluetooth label printer (M611).
Specifications
- Catalog No:
- 77282-372
- 77282-370
- 77282-376
- 77282-374
- 77282-368
- 77282-366
- 77282-364
- Connections:
- Mobile device connected, PC connected, and stand-alone
- Mobile device connected, PC connected, and stand-alone
- Mobile device connected, PC connected, and stand-alone
- Mobile device connected, PC connected, and stand-alone
- Mobile device connected, PC connected, and stand-alone
- Mobile device connected, PC connected, and stand-alone
- Mobile device connected, PC connected, and stand-alone
- Label size:
- 4" (max. label/tape width)
- 4" (max. label/tape width)
- 4" (max. label/tape width)
- 4" (max. label/tape width)
- 4" (max. label/tape width)
- 4" (max. label/tape width)
- 4" (max. label/tape width)
- Print resolution:
- 300 dpi
- 300 dpi
- 300 dpi
- 300 dpi
- 300 dpi
- 300 dpi
- 300 dpi
- Print speed:
- 5"/s (max.)
- 5"/s (max.)
- 5"/s (max.)
- 5"/s (max.)
- 5"/s (max.)
- 5"/s (max.)
- 5"/s (max.)
- Width:
- 19"
- 19"
- 19"
- 19"
- 19"
- 19"
- 19"
- Depth:
- 14.25"
- 14.25"
- 14.25"
- 14.25"
- 14.25"
- 14.25"
- 14.25"
- Height:
- 11"
- 11"
- 11"
- 11"
- 11"
- 11"
- 11"