About this item
Vogel's test is a classic protocol where rodent subjects are water deprived and then placed in the an apparatus, while simultaneously exposed to punishment in the shape of a mild shock whenever fluid is retrieved.
- Smooth DC shock delivery
- Output current: 0 to 4 m, adjustable in 0.1 mA increments
- Voltage: 163 V (direct current)
- Easy to clean and remove fecal and urine chamber
- Interchangable colors and plates
The Vogel's test is used for measurement of the ability of drugs to alter the drinking behavior in these water deprived rodents. The test features an automated liquid pattern dispenser, optical detection of lick, adjustable liquid volume, lick sensitivity, and automated data collection.