Skilled Forelimb Tests
Supplier: MAZE ENGINEERS
Certificates
About this item
The skilled forelimb reaching task is a widely used motor assessment for rodents.
- Acrylic construction
- Non-reflective surfaces
Rewards include pellets and carbohydrates in the form of pasta, or a lever press for operant experiments.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.