Specifications
- Ø:4.76 mm
- Length:11 cm
- Description:Drosophila gravitaxis maze
- Material:Acrylic
Specifications
About this item
The Drosophila gravitaxis maze is used to observe positive or negative gravitaxis movement in Drosophila species.
- Acrylic construction
- Easy to clean
Drosophila navigate the maze by choosing either to walk upward or downward to choose their preferred exit. The number of up or down choices made by the flies in the maze is used to deduce the gravitaxis behavior of Drosophila.