About this item
The visual X maze, or visual-stimuli 4-arm maze (ViS4M), is used to quantitatively study visual and cognitive abilities in rodents.
- Acrylic construction, easy to clean
- Color and contrast modes
- Object recognition can be studied by placing contrasting objects
- Illuminance options of each arm include: low, medium, high, red high, equal
- Illuminance settings obtained from the red LED source
- Optional traning shock grids can be included
This X-shaped apparatus can be equipped with either light-emitting diodes (LEDs) to study disorders such as early functional decline associated with aging and Alzheimer’s disease progression, or to study color vision or contrast vision in rodents. The Visual X Maze (ViS4M) apparatus is composed of black acrylic walls attached to a glass or acrylic base. Transparent and semi-transparent white floor plates are placed on the bottom level of each arm of the maze using small acrylic track brackets.