Natural Water Dechlorinator
Catalog #: 470353-526
Supplier: AQUASPROUTS, INC
New Product
Specifications
- Description:Natural water dechlorinator
- Cat. no.:470353-526
Specifications
About this item
Aquaponic and Hydroponic safe water dechlorinator to breaks the chlorine/ammonia bond in chloramines.
- Made with Aloe Vera to enhance fish’s natural protective slime coat
- Non-toxic, food safe
- Will not interfere with biological filters
Natural, food-safe dechlorinator uses vitamin C to neutralize chlorine and chloramines. Non-toxic to fish, invertebrates, live plants, and aquatic microbes, as well as humans and pets. Formulated using natural ingredients without noxious sulfur compounds.
Packaging: 4 fl. oz. bottle treats 240 gallons.
Customers Who Bought This Also Bought