About this item
Mouse monoclonal [38F3] antibody to Fibrillarin for WB, ICC/IF, IHC and Flow Cytometry with samples derived from Human, Rat, Mouse, Drosophila, Caenorhabditis elegans, Saccharomyces cerevisiae, Zebrafish and Yeast.
- Validated Applications: WB, ICC/IF, IHC, Flow Cytometry
- Recommended Dilutions: WB: 1:100 to 1:500, ICC/IF: 1:10 to 1:500, IHC: 1:10 to 1:50
Type: Primary
Antigen: Fibrillarin
Clonality: Monoclonal
Clone: 38F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Rat;Mouse;Drosophila;Caenorhabditis elegans;Saccharomyces cerevisiae;Zebrafish;Yeast
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77063-490
- 77061-738
- Antigen symbol:
- Fibrillarin
- Fibrillarin
- Antigen name:
- 34 kD nucleolar scleroderma antigen
- 34 kD nucleolar scleroderma antigen
- Antigen synonyms:
- FIB|FIB1|Fibrillarin|FBRL_HUMAN|Fbl|Histone-glutamine methyltransferase|FLRN|34 kDa nucleolar scleroderma antigen|Nop1p
- FIB|FIB1|Fibrillarin|FBRL_HUMAN|Fbl|Histone-glutamine methyltransferase|FLRN|34 kDa nucleolar scleroderma antigen|Nop1p
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 38F3
- 38F3
- Reactivity:
- Zebrafish, Yeast, S. cerevisiae, Drosophila, C. elegans, Human, Rat, Mouse
- Zebrafish, Yeast, S. cerevisiae, Drosophila, C. elegans, Human, Rat, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- UniprotID# P15646
- UniprotID# P15646
- Isotype:
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Yeast nuclear preparation
- Yeast nuclear preparation
- Form:
- Liquid
- Liquid
- Molecular weight:
- 35 kDa
- 35 kDa
- Storage buffer:
- Supplied as an aliquot of concentrated hybridoma cell culture media with 5 mM sodium azide
- Supplied as an aliquot of concentrated hybridoma cell culture media with 5 mM sodium azide
- Storage temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Shipping temperature:
- Shipped on blue ice at +4 °C
- Shipped on blue ice at +4 °C
- Tested applications:
- ICC/IF, IHC
- ICC/IF, IHC