Anti-NR3B Goat Polyclonal Antibody
Catalog # 76961-702
Supplier: ANTIBODIES.COM LLC
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:GluN3B
- Antigen symbol:NR3B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Antigen synonyms:Glutamate [NMDA] receptor subunit 3B|NMDAR3B|NMDA type glutamate receptor subunit NR3B|Glutamate receptor ionotropic NMDA 3B|NMDA receptor subunit 3B|N-methyl-D-aspartate receptor subtype 3B|NMD3B_HUMAN|GRIN3B|Glutamate receptor ionotropic N methyl D aspartate 3B
- Concentration:100 µg at 0.5 mg/ml
- Immunogen:Synthetic peptide corresponding to Human GRIN3B (C terminal)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Sequence:C-EAPPHSGRPGSQ
- Storage buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C
- Form:Liquid
- Cat. no.:76961-702
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to NR3B for ELISA and WB with samples derived from Human.
- Validated Applications: ELISA, W
Type: Primary
Antigen: NR3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Frequently Bought Together