To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:ARSA
- Clonality:Polyclonal
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:ASNA1
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 µg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:Arsenical pump driving ATPase|ARSA 1|ARSA-I|ARSA I|ArsA arsenite transporter ATP binding homolog 1|ARSA1|ArsA arsenite transporter, ATP-binding, homolog 1 (bacterial)|ArsA arsenite transporter, ATP binding, E. coli, homolog of, 1|ARSAI
- Storage buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-PHEVRGADKVNT
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide corresponding to Human ASNA1 (internal region)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to ASNA1 for ELISA and WB with samples derived from Human, Mouse and Rat.
- Validated Applications: ELISA, W
Type: Primary
Antigen: ASNA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human;Mouse;Rat