About this item
Rabbit monoclonal [ARC51683] antibody to CD20 for WB, IHC, ICC/IF and Flow Cytometry with samples derived from Human.
- Validated Applications: WB, IHC, ICC/IF, Flow Cytometry
- Recommended Dilutions: WB: 1:500 to 1:1,000, IHC: 1:50 to 1:200, ICC/IF: 1:500 to 1:1,000, Flow Cytometry: 1:50 to 1:200
Type: Primary
Antigen: CD20
Clonality: Monoclonal
Clone: ARC51683
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77222-778
- Antigen name:
- APY
- Antibody type:
- Primary
- Clonality:
- Monoclonal
- Clone:
- ARC51683
- Host:
- Rabbit
- Gene ID:
- UniprotID# P11836
- Isotype:
- IgG