To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:EPND
- Clonality:Monoclonal
- Clone:S327A-38
- Gene ID:UniprotID# Q00959
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human,Rat,Mouse
- Antigen symbol:NMDAR2A
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 µg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:hNR2A|LKS|GRIN2A|GluN2A|GRIN 2A|Glutamate [NMDA] receptor subunit epsilon-1|FESD|Glutamate receptor|Glutamate receptor ionotropic N methyl D aspartate 2A
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7.4, with 50% Glycerol and 0.09% Sodium Azide
- Molecular weight:~170 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Fusion protein amino acids 75-325 (extracellular N-terminus) of rat GluN2A/NR2A.
- Tested applications:IHC, ICC/IF
- Purification:Protein G purification
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [S327A-38] antibody to NMDAR2A for WB, IHC and ICC/IF with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC, ICC/IF
- Recommended Dilutions: WB: 1:1,000, ICC/IF: 1:100
Type: Primary
Antigen: NMDAR2A
Clonality: Monoclonal
Clone: S327A-38
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human;Mouse;Rat