To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:GluN1
- Clonality:Monoclonal
- Clone:S308-48
- Gene ID:UniprotID# P35439
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Antigen symbol:NMDAR1
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 µg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:N methyl D aspartate receptor subunit NR1|MRD8|N methyl D aspartate receptor channel subunit zeta 1|Glutamate [NMDA] receptor subunit zeta-1|N methyl D aspartate receptor|glutamate receptor ionotropic, NMDA 1|Glutamate receptor ionotropic, N-methyl-D aspartate, subunit 1|Grin1|Glutamate receptor ionotropic N methyl D aspartate 1
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7.4, with 50% Glycerol and 0.09% Sodium Azide
- Molecular weight:~105 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Fusion protein corresponding to the extracellular N-terminus, amino acids 42-361, of rat NR1 protein.
- Tested applications:IHC, ICC/IF
- Purification:Protein G purification
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [S308-48] antibody to NMDAR1 for WB, IHC and ICC/IF with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC, ICC/IF
- Recommended Dilutions: WB: 1:1,000, IHC: 1:1,000, ICC/IF: 1:100
Type: Primary
Antigen: NMDAR1
Clonality: Monoclonal
Clone: S308-48
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse;Rat